The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308185.1 | NP_001295114.1 | 1 - 2 | 971 | 117 |
NM_020307.3 | NP_064703.1 | 1 - 2 | 971 | 117 | |
XM_005247648.3 | XP_005247705.1 | 1 - 2 | 355 | 117 | |
XM_006713710.3 | XP_006713773.1 | 1 - 2 | 415 | 117 | |
XM_011513011.2 | XP_011511313.1 | 1 - 2 | 338 | 117 | |
XM_017006891.1 | XP_016862380.1 | 1 - 2 | 386 | 117 | |
XM_017006892.1 | XP_016862381.1 | 1 - 2 | 403 | 117 | |
XM_017006893.1 | XP_016862382.1 | 1 - 2 | 386 | 117 | |
XM_017006894.1 | XP_016862383.1 | 1 - 2 | 403 | 117 | |
XM_017006895.1 | XP_016862384.1 | 1 - 2 | 355 | 117 | |
XM_017006896.1 | XP_016862385.1 | 1 - 2 | 355 | 117 | |
XM_017006897.1 | XP_016862386.1 | 1 - 2 | 355 | 117 | |
GenBank mRNA | AF180920.1 | - | 1 - 2 | 359 | 117 |
AF367476.1 | - | 1 - 2 | 305 | 117 | |
AF367477.1 | - | 1 - 2 | 305 | 117 | |
AK022974.1 | - | 1 - 2 | 358 | 117 | |
AK122738.1 | - | 1 - 2 | 971 | 117 | |
BC007081.1 | - | 1 - 2 | 358 | 117 | |
BC038394.1 | - | 1 - 2 | 346 | 117 | |
BC067812.1 | - | 1 - 2 | 347 | 117 |