The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308185.1 | NP_001295114.1 | 3 - 4 | 1161 | 100 |
NM_020307.3 | NP_064703.1 | 3 - 4 | 1161 | 100 | |
XM_006713710.3 | XP_006713773.1 | 3 - 4 | 605 | 100 | |
XM_017006896.1 | XP_016862385.1 | 3 - 4 | 545 | 100 | |
GenBank mRNA | AF180920.1 | - | 3 - 4 | 549 | 100 |
AF367476.1 | - | 3 - 4 | 495 | 100 | |
AK022974.1 | - | 3 - 4 | 548 | 100 | |
AK122738.1 | - | 3 - 4 | 1161 | 100 | |
BC007081.1 | - | 3 - 4 | 548 | 100 | |
BC067812.1 | - | 3 - 4 | 537 | 100 |