The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310.3 | NP_001301.1 | 1 - 2 | 358 | 58 |
GenBank mRNA | AF039081.1 | - | 1 - 2 | 294 | 58 |
AK307202.1 | - | 2 - 3 | 234 | 58 | |
BC104873.1 | - | 1 - 2 | 202 | 58 | |
BC106052.1 | - | 1 - 2 | 299 | 58 | |
BC112157.1 | - | 1 - 2 | 202 | 58 | |
BY795169.2 | - | 1 - 2 | 358 | 58 |