The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014329.4 | NP_055144.3 | 12 - 13 | 1706 | 73 |
GenBank mRNA | AK128582.1 | - | 11 - 12 | 2180 | 73 |
AK291049.1 | - | 12 - 13 | 1474 | 73 | |
AK302563.1 | - | 12 - 13 | 1472 | 73 | |
BC043616.1 | - | 12 - 13 | 1609 | 73 | |
BC053598.1 | - | 7 - 8 | 846 | 73 | |
BC064567.1 | - | 12 - 13 | 1603 | 73 | |
L26339.1 | - | 12 - 13 | 1603 | 73 | |
U17474.1 | - | 2 - 3 | 243 | 73 |