The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014329.4 | NP_055144.3 | 24 - 25 | 3521 | 81 |
GenBank mRNA | AK128582.1 | - | 23 - 24 | 3995 | 81 |
AK291049.1 | - | 24 - 25 | 3289 | 81 | |
BC043616.1 | - | 24 - 25 | 3424 | 81 | |
BC053598.1 | - | 19 - 20 | 2661 | 81 | |
BC064567.1 | - | 24 - 25 | 3418 | 81 | |
L26339.1 | - | 24 - 25 | 3418 | 81 | |
U17474.1 | - | 14 - 15 | 2058 | 81 |