Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR52E8
Entrez Gene ID
390079
Gene Name
olfactory receptor family 52 subfamily E member 8
Gene Aliases
OR11-54
Chromosome Location
Chr.11: 5856749 - 5857702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005168.1 NP_001005168.1 1 - 1 71 105
GenBank mRNA BC137030.1 - 1 - 1 103 105
BC137041.1 - 1 - 1 103 105

View Full Product Details