The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307936.1 | NP_001294865.1 | 10 - 11 | 1282 | 109 |
NM_018976.4 | NP_061849.2 | 11 - 12 | 1398 | 109 | |
GenBank mRNA | AB037803.2 | - | 10 - 11 | 958 | 109 |
AF259799.1 | - | 10 - 11 | 978 | 109 | |
AF298897.1 | - | 11 - 12 | 1309 | 109 | |
AJ344099.1 | - | 11 - 12 | 1310 | 109 | |
AK001700.1 | - | 10 - 11 | 1191 | 109 | |
AK172784.1 | - | 4 - 5 | 405 | 109 | |
AK291644.1 | - | 11 - 12 | 1102 | 109 | |
AK293858.1 | - | 4 - 5 | 513 | 109 | |
BC040342.1 | - | 11 - 12 | 1300 | 109 | |
CR457267.1 | - | 7 - 8 | 658 | 109 |