Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR64
Entrez Gene ID
128025
Gene Name
WD repeat domain 64
Gene Aliases
-
Chromosome Location
Chr.1: 241652278 - 241802767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144625.4 NP_653226.4 22 - 23 2888 85
XM_006711736.3 XP_006711799.1 23 - 24 2918 85
XM_011544085.2 XP_011542387.1 20 - 21 2317 85
XM_011544086.2 XP_011542388.1 21 - 22 2319 85
XM_017000315.1 XP_016855804.1 23 - 24 2653 85
GenBank mRNA AK057540.1 - 10 - 11 1019 85
AK122927.1 - 22 - 23 2728 85
AY327404.1 - 12 - 13 1493 85

View Full Product Details