Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR64
Entrez Gene ID
128025
Gene Name
WD repeat domain 64
Gene Aliases
-
Chromosome Location
Chr.1: 241652278 - 241802767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144625.4 NP_653226.4 23 - 24 3070 100
XM_006711736.3 XP_006711799.1 24 - 25 3100 100
XM_011544085.2 XP_011542387.1 21 - 22 2499 100
XM_011544086.2 XP_011542388.1 22 - 23 2501 100
XM_017000315.1 XP_016855804.1 24 - 25 2835 100
GenBank mRNA AK057540.1 - 11 - 12 1201 100
AK122927.1 - 23 - 24 2910 100
AY327404.1 - 13 - 14 1675 100

View Full Product Details