The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144625.4 | NP_653226.4 | 13 - 14 | 1948 | 70 |
XM_006711736.3 | XP_006711799.1 | 14 - 15 | 1978 | 70 | |
XM_011544085.2 | XP_011542387.1 | 11 - 12 | 1377 | 70 | |
XM_011544086.2 | XP_011542388.1 | 12 - 13 | 1379 | 70 | |
XM_011544087.2 | XP_011542389.1 | 14 - 15 | 1978 | 70 | |
XM_011544091.1 | XP_011542393.1 | 14 - 15 | 1978 | 70 | |
XM_011544092.2 | XP_011542394.1 | 14 - 15 | 1978 | 70 | |
XM_017000315.1 | XP_016855804.1 | 14 - 15 | 1713 | 70 | |
GenBank mRNA | AK057540.1 | - | 1 - 2 | 79 | 70 |
AY327404.1 | - | 8 - 9 | 1054 | 70 |