The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184896.1 | NP_001171825.1 | 10 - 11 | 1494 | 79 |
NM_001184897.1 | NP_001171826.1 | 10 - 11 | 1645 | 79 | |
NM_001184898.1 | NP_001171827.1 | 10 - 11 | 1316 | 79 | |
NM_015107.2 | NP_055922.1 | 10 - 11 | 1645 | 79 | |
XM_005261996.1 | XP_005262053.1 | 11 - 12 | 1338 | 79 | |
XM_005261997.3 | XP_005262054.1 | 10 - 11 | 1943 | 79 | |
XM_005261999.1 | XP_005262056.1 | 10 - 11 | 1314 | 79 | |
XM_005262000.1 | XP_005262057.1 | 10 - 11 | 1607 | 79 | |
XM_011530778.1 | XP_011529080.1 | 10 - 11 | 1607 | 79 | |
XM_017029361.1 | XP_016884850.1 | 10 - 11 | 1893 | 79 | |
XM_017029362.1 | XP_016884851.1 | 10 - 11 | 1893 | 79 | |
GenBank mRNA | AB029034.1 | - | 10 - 11 | 1321 | 79 |
AK021696.1 | - | 9 - 10 | 1149 | 79 | |
AK304272.1 | - | 10 - 11 | 1467 | 79 | |
BC053861.1 | - | 10 - 11 | 1349 | 79 | |
CR933612.1 | - | 10 - 11 | 79 |