Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTR
Entrez Gene ID
4548
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
HMAG, MS, cblG
Chromosome Location
Chr.1: 236794304 - 236903981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000254.2 NP_000245.2 14 - 15 1752 94
NM_001291939.1 NP_001278868.1 14 - 15 1752 94
NM_001291940.1 NP_001278869.1 13 - 14 1639 94
XM_005273141.4 XP_005273198.1 14 - 15 1721 94
XM_006711770.2 XP_006711833.1 8 - 9 700 94
XM_011544194.2 XP_011542496.1 13 - 14 2514 94
XM_017001329.1 XP_016856818.1 13 - 14 2514 94
XM_017001330.1 XP_016856819.1 13 - 14 2514 94
GenBank mRNA AK299904.1 - 14 - 15 1752 94
AK307390.1 - 14 - 15 1741 94
BC130616.1 - 14 - 15 1351 94
BC136440.1 - 14 - 15 1351 94
BC144095.1 - 14 - 15 1351 94
BC144097.1 - 14 - 15 1351 94
BC144098.1 - 13 - 14 1238 94
BX641132.1 - 1 - 2 72 94
U71285.1 - 14 - 15 1392 94
U73338.1 - 14 - 15 1723 94
U75743.1 - 14 - 15 1615 94

View Full Product Details