Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTR
Entrez Gene ID
4548
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
HMAG, MS, cblG
Chromosome Location
Chr.1: 236794304 - 236903981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000254.2 NP_000245.2 17 - 18 2234 85
NM_001291939.1 NP_001278868.1 17 - 18 2234 85
NM_001291940.1 NP_001278869.1 16 - 17 2121 85
XM_005273141.4 XP_005273198.1 17 - 18 2203 85
XM_006711770.2 XP_006711833.1 11 - 12 1182 85
XM_011544194.2 XP_011542496.1 16 - 17 2996 85
XM_017001329.1 XP_016856818.1 16 - 17 2996 85
XM_017001330.1 XP_016856819.1 16 - 17 2996 85
GenBank mRNA AK307390.1 - 17 - 18 2223 85
BC130616.1 - 17 - 18 1833 85
BC136440.1 - 17 - 18 1833 85
BC144095.1 - 17 - 18 1833 85
BC144097.1 - 17 - 18 1833 85
BC144098.1 - 16 - 17 1720 85
BX641132.1 - 4 - 5 473 85
U71285.1 - 17 - 18 1874 85
U73338.1 - 17 - 18 2205 85
U75743.1 - 17 - 18 2097 85

View Full Product Details