Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PREX2
Entrez Gene ID
80243
Gene Name
phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2
Gene Aliases
DEP.2, DEPDC2, P-REX2, PPP1R129
Chromosome Location
Chr.8: 67951921 - 68237033 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024870.3 NP_079146.2 16 - 17 2069 193
NM_025170.5 NP_079446.3 16 - 17 2069 193
XM_011517612.2 XP_011515914.1 16 - 17 2263 193
GenBank mRNA AJ437636.2 - 16 - 17 2066 193
AK023049.1 - 11 - 12 1144 193
AK024079.1 - 1 - 2 145 193
AK294299.1 - 14 - 15 1500 193
AY508996.1 - 16 - 17 1816 193
AY508997.1 - 16 - 17 1816 193

View Full Product Details