The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002894.2 | NP_002885.1 | 6 - 7 | 754 | 133 |
NM_203291.1 | NP_976036.1 | 6 - 7 | 724 | 133 | |
NM_203292.1 | NP_976037.1 | 6 - 7 | 805 | 133 | |
XM_005258325.2 | XP_005258382.1 | 6 - 7 | 685 | 133 | |
XM_005258326.3 | XP_005258383.1 | 5 - 6 | 521 | 133 | |
XM_006722519.2 | XP_006722582.1 | 8 - 9 | 826 | 133 | |
XM_006722520.2 | XP_006722583.1 | 7 - 8 | 762 | 133 | |
XM_006722521.2 | XP_006722584.1 | 6 - 7 | 822 | 133 | |
XM_011526132.2 | XP_011524434.1 | 7 - 8 | 773 | 133 | |
XM_017025915.1 | XP_016881404.1 | 4 - 5 | 304 | 133 | |
XM_017025916.1 | XP_016881405.1 | 7 - 8 | 751 | 133 | |
GenBank mRNA | AF043431.1 | - | 6 - 7 | 721 | 133 |
AK292481.1 | - | 6 - 7 | 685 | 133 | |
BC001170.1 | - | 6 - 7 | 910 | 133 | |
BC030590.1 | - | 6 - 7 | 552 | 133 | |
BU187672.1 | - | 6 - 7 | 694 | 133 | |
BX648221.1 | - | 6 - 7 | 779 | 133 | |
U72066.1 | - | 6 - 7 | 722 | 133 |