The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003875.2 | NP_003866.1 | 4 - 5 | 758 | 135 |
XM_011513263.2 | XP_011511565.1 | 4 - 5 | 456 | 135 | |
XM_017007422.1 | XP_016862911.1 | 6 - 7 | 943 | 135 | |
GenBank mRNA | AK223399.1 | - | 4 - 5 | 641 | 135 |
AK291504.1 | - | 4 - 5 | 626 | 135 | |
AK300787.1 | - | 5 - 6 | 522 | 135 | |
AK302148.1 | - | 2 - 3 | 347 | 135 | |
AK315832.1 | - | 5 - 6 | 533 | 135 | |
BC012178.1 | - | 4 - 5 | 626 | 135 | |
U10860.1 | - | 4 - 5 | 545 | 135 |