The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003875.2 | NP_003866.1 | 6 - 7 | 1054 | 112 |
XM_011513263.2 | XP_011511565.1 | 6 - 7 | 752 | 112 | |
XM_017007422.1 | XP_016862911.1 | 8 - 9 | 1239 | 112 | |
GenBank mRNA | AK223399.1 | - | 6 - 7 | 937 | 112 |
AK291504.1 | - | 6 - 7 | 922 | 112 | |
AK300787.1 | - | 7 - 8 | 818 | 112 | |
AK302148.1 | - | 4 - 5 | 643 | 112 | |
AK315832.1 | - | 7 - 8 | 829 | 112 | |
BC012178.1 | - | 6 - 7 | 922 | 112 | |
U10860.1 | - | 6 - 7 | 841 | 112 |