The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003875.2 | NP_003866.1 | 8 - 9 | 1376 | 98 |
XM_011513263.2 | XP_011511565.1 | 8 - 9 | 1074 | 98 | |
XM_017007422.1 | XP_016862911.1 | 10 - 11 | 1561 | 98 | |
GenBank mRNA | AK223399.1 | - | 8 - 9 | 1259 | 98 |
AK291504.1 | - | 8 - 9 | 1244 | 98 | |
AK300787.1 | - | 9 - 10 | 1140 | 98 | |
AK302148.1 | - | 6 - 7 | 965 | 98 | |
AK315832.1 | - | 9 - 10 | 1151 | 98 | |
BC012178.1 | - | 8 - 9 | 1244 | 98 | |
U10860.1 | - | 8 - 9 | 1163 | 98 |