Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.19: 4182618 - 4224814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133475.1 NP_597732.1 21 - 22 3636 62
XM_011527756.2 XP_011526058.1 23 - 24 3741 62
XM_011527757.2 XP_011526059.1 21 - 22 3794 62
XM_011527758.2 XP_011526060.1 22 - 23 3654 62
XM_011527761.1 XP_011526063.1 24 - 25 3811 62
XM_011527764.2 XP_011526066.1 20 - 21 3707 62
XM_011527765.2 XP_011526067.1 21 - 22 3555 62
XM_011527768.2 XP_011526070.1 9 - 10 2512 62
XM_017026438.1 XP_016881927.1 24 - 25 3802 62
XM_017026439.1 XP_016881928.1 23 - 24 3724 62
XM_017026440.1 XP_016881929.1 22 - 23 3697 62
XM_017026441.1 XP_016881930.1 12 - 13 2839 62

View Full Product Details