Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.19: 4182618 - 4224814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133475.1 NP_597732.1 5 - 6 614 81
XM_011527756.2 XP_011526058.1 5 - 6 497 81
XM_011527757.2 XP_011526059.1 5 - 6 772 81
XM_011527758.2 XP_011526060.1 4 - 5 410 81
XM_011527761.1 XP_011526063.1 6 - 7 567 81
XM_011527764.2 XP_011526066.1 4 - 5 685 81
XM_011527765.2 XP_011526067.1 3 - 4 311 81
XM_017026438.1 XP_016881927.1 6 - 7 558 81
XM_017026439.1 XP_016881928.1 5 - 6 480 81
XM_017026440.1 XP_016881929.1 5 - 6 480 81
GenBank mRNA AB075861.1 - 3 - 4 81
BC028236.1 - 3 - 4 81
BI914435.1 - 3 - 4 339 81

View Full Product Details