The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133475.1 | NP_597732.1 | 9 - 10 | 923 | 81 |
XM_011527756.2 | XP_011526058.1 | 10 - 11 | 833 | 81 | |
XM_011527757.2 | XP_011526059.1 | 9 - 10 | 1081 | 81 | |
XM_011527758.2 | XP_011526060.1 | 9 - 10 | 746 | 81 | |
XM_011527761.1 | XP_011526063.1 | 11 - 12 | 903 | 81 | |
XM_011527764.2 | XP_011526066.1 | 8 - 9 | 994 | 81 | |
XM_011527765.2 | XP_011526067.1 | 8 - 9 | 647 | 81 | |
XM_017026438.1 | XP_016881927.1 | 11 - 12 | 894 | 81 | |
XM_017026439.1 | XP_016881928.1 | 10 - 11 | 816 | 81 | |
XM_017026440.1 | XP_016881929.1 | 9 - 10 | 789 | 81 | |
GenBank mRNA | AB075861.1 | - | 7 - 8 | 620 | 81 |
BC028236.1 | - | 7 - 8 | 565 | 81 |