The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198274.3 | NP_938015.1 | 6 - 7 | 970 | 140 |
XM_005264156.2 | XP_005264213.1 | 5 - 6 | 886 | 140 | |
GenBank mRNA | AK091724.1 | - | 6 - 7 | 888 | 140 |
AL832035.1 | - | 6 - 7 | 887 | 140 | |
AY518933.1 | - | 6 - 7 | 867 | 140 | |
AY518934.1 | - | 2 - 3 | 236 | 140 | |
BC126191.1 | - | 6 - 7 | 970 | 140 | |
BX647878.1 | - | 6 - 7 | 889 | 140 |