The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017921.3 | NP_060391.2 | 16 - 17 | 1895 | 102 |
NR_130139.1 | 5 - 6 | 467 | 102 | ||
NR_130140.1 | 2 - 3 | 271 | 102 | ||
XM_011524981.1 | XP_011523283.1 | 16 - 17 | 1905 | 102 | |
GenBank mRNA | AK023046.1 | - | 14 - 15 | 1531 | 102 |
AK023272.1 | - | 13 - 14 | 1474 | 102 | |
AK074322.1 | - | 2 - 3 | 167 | 102 | |
AL834300.1 | - | 13 - 14 | 1407 | 102 | |
BC025930.1 | - | 16 - 17 | 1836 | 102 | |
BU542911.1 | - | 5 - 6 | 435 | 102 | |
DA998574.1 | - | 2 - 3 | 271 | 102 |