The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017921.3 | NP_060391.2 | 1 - 2 | 235 | 76 |
XM_011524979.1 | XP_011523281.1 | 1 - 2 | 230 | 76 | |
XM_011524980.1 | XP_011523282.1 | 1 - 2 | 230 | 76 | |
XM_011524981.1 | XP_011523283.1 | 1 - 2 | 230 | 76 | |
XM_011524982.2 | XP_011523284.1 | 1 - 2 | 230 | 76 | |
GenBank mRNA | AB040932.1 | - | 1 - 2 | 142 | 76 |
AK000664.1 | - | 1 - 2 | 144 | 76 | |
AK294473.1 | - | 1 - 2 | 161 | 76 | |
BC025930.1 | - | 1 - 2 | 176 | 76 | |
DC322260.1 | - | 1 - 2 | 194 | 76 | |
HY116973.1 | - | 1 - 2 | 237 | 76 |