Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr.17: 81539885 - 81552403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033338.1 2 - 3 554 138
XM_017025162.1 XP_016880651.1 2 - 3 379 138
GenBank mRNA BC008883.2 - 2 - 3 527 138

View Full Product Details