Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr.17: 81539885 - 81552403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025161.5 NP_079437.5 3 - 4 1322 100
NR_033338.1 3 - 4 1518 100
XM_006722111.2 XP_006722174.1 3 - 4 1317 100
XM_006722112.3 XP_006722175.1 3 - 4 1468 100
XM_017025162.1 XP_016880651.1 3 - 4 1343 100
XM_017025163.1 XP_016880652.1 3 - 4 1239 100
GenBank mRNA BC008883.2 - 3 - 4 1491 100
BC117139.1 - 1 - 2 842 100
BC117141.1 - 1 - 2 842 100
BC143861.1 - 1 - 2 842 100
DQ989324.1 - 3 - 4 1304 100

View Full Product Details