The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289082.1 | NP_001276011.1 | 3 - 4 | 691 | 115 |
NM_001289083.1 | NP_001276012.1 | 2 - 3 | 517 | 115 | |
NM_001289085.1 | NP_001276014.1 | 3 - 4 | 691 | 115 | |
NM_001289087.1 | NP_001276016.1 | 2 - 3 | 517 | 115 | |
NM_139018.4 | NP_620587.2 | 2 - 3 | 517 | 115 | |
NR_110298.1 | 3 - 4 | 691 | 115 | ||
XM_017024215.1 | XP_016879704.1 | 2 - 3 | 517 | 115 | |
GenBank mRNA | AF251706.1 | - | 2 - 3 | 389 | 115 |
AF375480.1 | - | 2 - 3 | 447 | 115 | |
AK092757.1 | - | 2 - 3 | 471 | 115 | |
AK315165.1 | - | 2 - 3 | 440 | 115 | |
AY358545.1 | - | 2 - 3 | 464 | 115 | |
BC028199.1 | - | 3 - 4 | 669 | 115 | |
BI911496.1 | - | 2 - 3 | 413 | 115 | |
BM922663.1 | - | 2 - 3 | 386 | 115 | |
BP226373.1 | - | 2 - 3 | 440 | 115 |