Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUVBL2
Entrez Gene ID
10856
Gene Name
RuvB like AAA ATPase 2
Gene Aliases
CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B
Chromosome Location
Chr.19: 48993448 - 49015995 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321190.1 NP_001308119.1 8 - 9 1200 67
NM_001321191.1 NP_001308120.1 8 - 9 1109 67
NM_006666.2 NP_006657.1 8 - 9 1130 67
NR_135578.1 8 - 9 1119 67
XM_011526330.1 XP_011524632.1 8 - 9 1043 67
GenBank mRNA AB024301.1 - 8 - 9 679 67
AF124607.1 - 8 - 9 687 67
AF155138.1 - 8 - 9 672 67
AK027762.1 - 8 - 9 750 67
AK057498.1 - 8 - 9 1677 67
AK074542.1 - 8 - 9 680 67
AL136743.1 - 7 - 8 669 67
BC000428.2 - 8 - 9 671 67
BC004531.2 - 8 - 9 679 67
BC008355.1 - 8 - 9 668 67
CR533507.1 - 7 - 8 666 67
Y18417.1 - 8 - 9 671 67

View Full Product Details