The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002944.2 | NP_002935.2 | 12 - 13 | 1956 | 100 |
XM_011536049.2 | XP_011534351.1 | 13 - 14 | 2091 | 100 | |
XM_011536050.2 | XP_011534352.1 | 13 - 14 | 2091 | 100 | |
XM_011536051.2 | XP_011534353.1 | 12 - 13 | 2064 | 100 | |
XM_011536054.2 | XP_011534356.1 | 13 - 14 | 2092 | 100 | |
XM_011536055.2 | XP_011534357.1 | 13 - 14 | 2091 | 100 | |
XM_011536056.2 | XP_011534358.1 | 13 - 14 | 2090 | 100 | |
XM_011536057.2 | XP_011534359.1 | 13 - 14 | 2089 | 100 | |
XM_011536058.2 | XP_011534360.1 | 13 - 14 | 2089 | 100 | |
GenBank mRNA | M34353.1 | - | 12 - 13 | 1963 | 100 |