The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002944.2 | NP_002935.2 | 17 - 18 | 2799 | 68 |
XM_006715548.3 | XP_006715611.1 | 18 - 19 | 2891 | 68 | |
XM_011536050.2 | XP_011534352.1 | 18 - 19 | 2934 | 68 | |
XM_017011173.1 | XP_016866662.1 | 17 - 18 | 2967 | 68 | |
GenBank mRNA | BC144556.1 | - | 17 - 18 | 2391 | 68 |
M34353.1 | - | 17 - 18 | 2806 | 68 |