The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002944.2 | NP_002935.2 | 1 - 2 | 326 | 59 |
XM_006715548.3 | XP_006715611.1 | 1 - 2 | 433 | 59 | |
XM_011536049.2 | XP_011534351.1 | 1 - 2 | 434 | 59 | |
XM_011536050.2 | XP_011534352.1 | 1 - 2 | 434 | 59 | |
XM_011536051.2 | XP_011534353.1 | 1 - 2 | 434 | 59 | |
XM_011536052.2 | XP_011534354.1 | 1 - 2 | 434 | 59 | |
XM_011536053.2 | XP_011534355.1 | 1 - 2 | 434 | 59 | |
XM_011536054.2 | XP_011534356.1 | 1 - 2 | 435 | 59 | |
XM_011536055.2 | XP_011534357.1 | 1 - 2 | 434 | 59 | |
XM_011536056.2 | XP_011534358.1 | 1 - 2 | 433 | 59 | |
XM_011536057.2 | XP_011534359.1 | 1 - 2 | 432 | 59 | |
XM_011536058.2 | XP_011534360.1 | 1 - 2 | 432 | 59 | |
XM_017011172.1 | XP_016866661.1 | 1 - 2 | 434 | 59 | |
XM_017011173.1 | XP_016866662.1 | 1 - 2 | 536 | 59 | |
GenBank mRNA | BC144556.1 | - | 1 - 2 | 174 | 59 |
M34353.1 | - | 1 - 2 | 333 | 59 |