The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242816.1 | NP_001229745.1 | 4 - 5 | 417 | 62 |
GenBank mRNA | AK131370.1 | - | 4 - 5 | 353 | 62 |