The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008800.2 | NP_001008800.1 | 10 - 11 | 1523 | 65 |
NM_005998.4 | NP_005989.3 | 12 - 13 | 1637 | 65 | |
NR_036564.1 | 13 - 14 | 1799 | 65 | ||
NR_036565.1 | 13 - 14 | 1750 | 65 | ||
GenBank mRNA | AB208882.1 | - | 12 - 13 | 1503 | 65 |
AK126400.1 | - | 12 - 13 | 1424 | 65 | |
AK293477.1 | - | 10 - 11 | 1353 | 65 | |
AK300765.1 | - | 13 - 14 | 1631 | 65 | |
AK315923.1 | - | 11 - 12 | 1423 | 65 | |
AK316387.1 | - | 13 - 14 | 1542 | 65 | |
AL833197.1 | - | 12 - 13 | 1497 | 65 | |
AY524046.1 | - | 12 - 13 | 1462 | 65 | |
BC006501.2 | - | 12 - 13 | 1469 | 65 | |
BC008019.1 | - | 12 - 13 | 1479 | 65 | |
U17104.1 | - | 2 - 3 | 376 | 65 | |
X74801.1 | - | 12 - 13 | 1403 | 65 |