The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008800.2 | NP_001008800.1 | 7 - 8 | 1003 | 71 |
NM_005998.4 | NP_005989.3 | 9 - 10 | 1117 | 71 | |
NR_036564.1 | 10 - 11 | 1279 | 71 | ||
NR_036565.1 | 10 - 11 | 1230 | 71 | ||
GenBank mRNA | AB208882.1 | - | 9 - 10 | 983 | 71 |
AK126400.1 | - | 9 - 10 | 904 | 71 | |
AK293477.1 | - | 7 - 8 | 833 | 71 | |
AK300765.1 | - | 10 - 11 | 1111 | 71 | |
AK308023.1 | - | 8 - 9 | 809 | 71 | |
AK315923.1 | - | 8 - 9 | 903 | 71 | |
AK316387.1 | - | 10 - 11 | 1022 | 71 | |
AL833197.1 | - | 9 - 10 | 977 | 71 | |
AY524046.1 | - | 9 - 10 | 942 | 71 | |
BC006501.2 | - | 9 - 10 | 949 | 71 | |
BC008019.1 | - | 9 - 10 | 959 | 71 | |
X74801.1 | - | 9 - 10 | 883 | 71 |