Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCC11
Entrez Gene ID
85320
Gene Name
ATP binding cassette subfamily C member 11
Gene Aliases
EWWD, MRP8, WW
Chromosome Location
Chr.16: 48164842 - 48249973 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032583.3 NP_115972.2 27 - 28 3843 72
NM_033151.3 NP_149163.2 26 - 27 4129 72
XM_011523397.1 XP_011521699.1 23 - 24 3349 72
XM_011523398.2 XP_011521700.1 15 - 16 2064 72
XM_017023795.1 XP_016879284.1 27 - 28 6457 72
XM_017023796.1 XP_016879285.1 28 - 29 3930 72
XM_017023797.1 XP_016879286.1 28 - 29 4058 72
XM_017023798.1 XP_016879287.1 27 - 28 3884 72
XM_017023799.1 XP_016879288.1 27 - 28 3866 72
XM_017023800.1 XP_016879289.1 27 - 28 3857 72
XM_017023801.1 XP_016879290.1 26 - 27 6349 72
XM_017023802.1 XP_016879291.1 21 - 22 3204 72
GenBank mRNA AF352582.1 - 27 - 28 3878 72
AF367202.1 - 27 - 28 3857 72
AL117406.1 - 8 - 9 1079 72
AY040219.1 - 26 - 27 4129 72

View Full Product Details