Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHCBP1
Entrez Gene ID
79801
Gene Name
SHC binding and spindle associated 1
Gene Aliases
PAL
Chromosome Location
Chr.16: 46578591 - 46624933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324319.1 NP_001311248.1 2 - 3 313 71
NM_024745.4 NP_079021.3 2 - 3 310 71
NR_136738.1 2 - 3 313 71
XM_011523335.2 XP_011521637.1 2 - 3 3844 71
GenBank mRNA AK055931.1 - 2 - 3 281 71
AK292164.1 - 2 - 3 277 71
AK310430.1 - 2 - 3 279 71
AK315706.1 - 2 - 3 279 71
BC030699.1 - 2 - 3 285 71
BP245197.1 - 2 - 3 310 71

View Full Product Details