Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSCAN20
Entrez Gene ID
7579
Gene Name
zinc finger and SCAN domain containing 20
Gene Aliases
KOX29, ZFP-31, ZNF31, ZNF360
Chromosome Location
Chr.1: 33472591 - 33504253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145238.3 NP_660281.2 6 - 7 1612 63
XM_006710874.3 XP_006710937.1 6 - 7 1596 63
XM_006710876.3 XP_006710939.1 4 - 5 1434 63
XM_017002237.1 XP_016857726.1 6 - 7 1639 63
XM_017002239.1 XP_016857728.1 5 - 6 1579 63
XM_017002240.1 XP_016857729.1 5 - 6 1511 63
XM_017002241.1 XP_016857730.1 4 - 5 2082 63
GenBank mRNA AK131405.1 - 6 - 7 1598 63
BC008827.2 - 7 - 8 1391 63

View Full Product Details