The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286771.2 | NP_001273700.1 | 4 - 5 | 594 | 73 |
NM_015574.1 | NP_056389.1 | 4 - 5 | 976 | 73 | |
NM_032217.4 | NP_115593.3 | 4 - 5 | 976 | 73 | |
NM_198889.2 | NP_942592.1 | 4 - 5 | 976 | 73 | |
XM_005265671.3 | XP_005265728.1 | 4 - 5 | 1320 | 73 | |
XM_005265672.3 | XP_005265729.1 | 4 - 5 | 1320 | 73 | |
XM_005265673.3 | XP_005265730.1 | 4 - 5 | 1320 | 73 | |
XM_017008011.1 | XP_016863500.1 | 4 - 5 | 551 | 73 | |
XM_017008012.1 | XP_016863501.1 | 4 - 5 | 551 | 73 | |
XM_017008013.1 | XP_016863502.1 | 4 - 5 | 551 | 73 | |
XM_017008014.1 | XP_016863503.1 | 4 - 5 | 1320 | 73 | |
XM_017008015.1 | XP_016863504.1 | 4 - 5 | 551 | 73 | |
XM_017008016.1 | XP_016863505.1 | 4 - 5 | 1320 | 73 | |
XM_017008017.1 | XP_016863506.1 | 4 - 5 | 551 | 73 | |
GenBank mRNA | AB014597.2 | - | 4 - 5 | 500 | 73 |
AF308285.1 | - | 4 - 5 | 963 | 73 | |
AK300302.1 | - | 4 - 5 | 550 | 73 | |
AL833287.1 | - | 1 - 2 | 153 | 73 | |
BC004173.2 | - | 4 - 5 | 957 | 73 | |
BC007747.2 | - | 2 - 3 | 306 | 73 | |
BC043394.1 | - | 4 - 5 | 945 | 73 | |
BX648035.1 | - | 4 - 5 | 549 | 73 |