The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256370.1 | NP_001243299.1 | 6 - 7 | 940 | 82 |
NM_001286523.1 | NP_001273452.1 | 6 - 7 | 784 | 82 | |
NM_022136.4 | NP_071419.3 | 5 - 6 | 660 | 82 | |
XM_011529684.2 | XP_011527986.1 | 6 - 7 | 700 | 82 | |
XM_011529685.1 | XP_011527987.1 | 4 - 5 | 465 | 82 | |
XM_011529686.1 | XP_011527988.1 | 5 - 6 | 657 | 82 | |
XM_017028427.1 | XP_016883916.1 | 5 - 6 | 657 | 82 | |
GenBank mRNA | AF218085.2 | - | 6 - 7 | 940 | 82 |
AF222927.1 | - | 5 - 6 | 643 | 82 | |
AK125144.1 | - | 5 - 6 | 643 | 82 | |
BC029112.1 | - | 5 - 6 | 634 | 82 | |
HG380514.1 | - | 6 - 7 | 759 | 82 |