Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMSN1
Entrez Gene ID
64092
Gene Name
SAM domain, SH3 domain and nuclear localization signals 1
Gene Aliases
HACS1, NASH1, SASH2, SH3D6B, SLy2
Chromosome Location
Chr.21: 14485228 - 14583402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256370.1 NP_001243299.1 7 - 8 1144 146
NM_001286523.1 NP_001273452.1 7 - 8 988 146
NM_022136.4 NP_071419.3 6 - 7 864 146
XM_011529684.2 XP_011527986.1 7 - 8 904 146
XM_011529685.1 XP_011527987.1 5 - 6 669 146
XM_011529686.1 XP_011527988.1 6 - 7 861 146
XM_017028427.1 XP_016883916.1 6 - 7 861 146
GenBank mRNA AF218085.2 - 7 - 8 1144 146
AF222927.1 - 6 - 7 847 146
AF519621.1 - 2 - 3 342 146
AK125144.1 - 6 - 7 847 146
BC029112.1 - 6 - 7 838 146
HG380514.1 - 7 - 8 963 146

View Full Product Details