The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001927.3 | NP_001918.3 | 1 - 2 | 663 | 86 |
GenBank mRNA | AF055081.1 | - | 1 - 2 | 577 | 86 |
AF055082.1 | - | 1 - 2 | 577 | 86 | |
AF055083.1 | - | 1 - 2 | 577 | 86 | |
AF137053.1 | - | 1 - 2 | 577 | 86 | |
AF167579.1 | - | 1 - 2 | 577 | 86 | |
AF486807.1 | - | 1 - 2 | 577 | 86 | |
AF487828.1 | - | 1 - 2 | 577 | 86 | |
AF521879.1 | - | 1 - 2 | 577 | 86 | |
AF527578.1 | - | 1 - 2 | 577 | 86 | |
AK097038.1 | - | 2 - 3 | 603 | 86 | |
AK098332.1 | - | 2 - 3 | 457 | 86 | |
AK123787.1 | - | 2 - 3 | 402 | 86 | |
AK300654.1 | - | - | 658 | 86 | |
AY083345.1 | - | 1 - 2 | 577 | 86 | |
AY125465.1 | - | 1 - 2 | 577 | 86 | |
BC032116.2 | - | 1 - 2 | 675 | 86 | |
DQ104335.1 | - | 1 - 2 | 577 | 86 | |
DQ104336.1 | - | 1 - 2 | 577 | 86 | |
DQ104337.1 | - | 1 - 2 | 577 | 86 | |
EF617312.1 | - | 1 - 2 | 657 | 86 | |
JX114780.1 | - | 1 - 2 | 577 | 86 | |
U59167.1 | - | 1 - 2 | 654 | 86 |