The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199763.1 | NP_001186692.1 | 10 - 11 | 1757 | 68 |
NM_001199764.1 | NP_001186693.1 | 11 - 12 | 1614 | 68 | |
NM_002846.3 | NP_002837.1 | 11 - 12 | 1844 | 68 | |
XM_011511566.1 | XP_011509868.1 | 11 - 12 | 1693 | 68 | |
XM_017004609.1 | XP_016860098.1 | 11 - 12 | 1696 | 68 | |
XM_017004610.1 | XP_016860099.1 | 10 - 11 | 1609 | 68 | |
XM_017004611.1 | XP_016860100.1 | 11 - 12 | 1696 | 68 | |
XM_017004612.1 | XP_016860101.1 | 10 - 11 | 1606 | 68 | |
GenBank mRNA | AB209368.1 | - | 9 - 10 | 2077 | 68 |
AK122679.1 | - | 2 - 3 | 358 | 68 | |
AK296335.1 | - | 11 - 12 | 1591 | 68 | |
AK314574.1 | - | 10 - 11 | 1521 | 68 | |
BC007713.2 | - | 7 - 8 | 1099 | 68 | |
BC070053.1 | - | 10 - 11 | 1583 | 68 | |
BT006975.1 | - | 3 - 4 | 439 | 68 | |
L18983.1 | - | 11 - 12 | 1676 | 68 | |
X62899.2 | - | 9 - 10 | 1366 | 68 |