Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAMP
Entrez Gene ID
14
Gene Name
angio associated migratory cell protein
Gene Aliases
-
Chromosome Location
Chr.2: 218264129 - 218270209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001087.4 NP_001078.2 10 - 11 1351 102
NM_001302545.1 NP_001289474.1 10 - 11 1354 102
GenBank mRNA AB209790.1 - 7 - 8 1790 102
AK131047.1 - 10 - 11 1288 102
AK225078.1 - 10 - 11 1259 102
BC008809.2 - 7 - 8 834 102
BC020244.1 - 10 - 11 1215 102
BC039866.1 - 10 - 11 1279 102
CR456755.1 - 10 - 11 1282 102
M95627.1 - 10 - 11 1282 102

View Full Product Details