Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAMP
Entrez Gene ID
14
Gene Name
angio associated migratory cell protein
Gene Aliases
-
Chromosome Location
Chr.2: 218264129 - 218270209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001087.4 NP_001078.2 7 - 8 1007 66
NM_001302545.1 NP_001289474.1 7 - 8 1010 66
GenBank mRNA AK131047.1 - 7 - 8 944 66
AK225078.1 - 7 - 8 915 66
BC008809.2 - 4 - 5 490 66
BC020244.1 - 7 - 8 871 66
BC039866.1 - 7 - 8 935 66
CR456755.1 - 7 - 8 938 66
M95627.1 - 7 - 8 938 66

View Full Product Details