Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
Chromosome Location
Chr.2: 218270392 - 218346793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015488.4 NP_056303.3 7 - 8 929 62
NM_022572.4 NP_072094.1 6 - 7 802 62
GenBank mRNA AB033010.1 - 7 - 8 770 62
AK289867.1 - 7 - 8 837 62
AK309308.1 - - 632 62
AL137675.1 - 5 - 6 1428 62
AY358753.1 - 6 - 7 722 62
BC002937.2 - 6 - 7 713 62
BC036457.2 - 7 - 8 831 62

View Full Product Details