Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH2B1
Entrez Gene ID
25970
Gene Name
SH2B adaptor protein 1
Gene Aliases
PSM, SH2B
Chromosome Location
Chr.16: 28846600 - 28874213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145796.1 NP_001139268.1 8 - 9 2171 126
NM_001145812.1 NP_001139284.1 8 - 9 2270 126
NM_001308294.1 NP_001295223.1 7 - 8 1106 126
NM_015503.2 NP_056318.2 7 - 8 4001 126
GenBank mRNA AB037720.1 - 7 - 8 5190 126
AF227968.1 - 8 - 9 2027 126
AK027488.1 - 7 - 8 1336 126
AK290332.1 - 8 - 9 2270 126
AK297080.1 - 7 - 8 1057 126
AL713760.1 - 7 - 8 4001 126
BC010704.1 - 7 - 8 1724 126

View Full Product Details