Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
Chromosome Location
Chr.7: 91940867 - 92110673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005751.4 NP_005742.4 23 - 24 5985 139
NM_147185.2 NP_671714.1 23 - 24 5985 139
XM_017011642.1 XP_016867131.1 24 - 25 6089 139
XM_017011643.1 XP_016867132.1 24 - 25 6089 139
XM_017011644.1 XP_016867133.1 24 - 25 6089 139
XM_017011645.1 XP_016867134.1 23 - 24 6035 139
XM_017011646.1 XP_016867135.1 24 - 25 6089 139
XM_017011647.1 XP_016867136.1 23 - 24 5996 139
XM_017011648.1 XP_016867137.1 23 - 24 5993 139
XM_017011649.1 XP_016867138.1 24 - 25 6089 139
XM_017011650.1 XP_016867139.1 23 - 24 5993 139
XM_017011651.1 XP_016867140.1 23 - 24 5948 139
XM_017011652.1 XP_016867141.1 24 - 25 6089 139
XM_017011653.1 XP_016867142.1 22 - 23 5900 139
XM_017011654.1 XP_016867143.1 22 - 23 5852 139
XM_017011655.1 XP_016867144.1 24 - 25 5748 139
XM_017011656.1 XP_016867145.1 23 - 24 5678 139
XM_017011657.1 XP_016867146.1 9 - 10 1598 139
XM_017011658.1 XP_016867147.1 2 - 3 960 139
XM_017011659.1 XP_016867148.1 2 - 3 960 139
XM_017011660.1 XP_016867149.1 2 - 3 960 139
GenBank mRNA AB019691.1 - 23 - 24 5846 139
AF083037.1 - 16 - 17 4824 139
AF247727.4 - 22 - 23 5619 139
AJ131693.1 - 23 - 24 5985 139
AL117418.1 - 2 - 3 949 139
AY528715.1 - 5 - 6 832 139
BC062448.1 - 2 - 3 139

View Full Product Details