Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
Chromosome Location
Chr.7: 91940867 - 92110673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005751.4 NP_005742.4 5 - 6 804 92
NM_147185.2 NP_671714.1 5 - 6 804 92
XM_017011642.1 XP_016867131.1 5 - 6 812 92
XM_017011643.1 XP_016867132.1 5 - 6 812 92
XM_017011644.1 XP_016867133.1 5 - 6 812 92
XM_017011645.1 XP_016867134.1 4 - 5 758 92
XM_017011646.1 XP_016867135.1 5 - 6 812 92
XM_017011647.1 XP_016867136.1 5 - 6 812 92
XM_017011648.1 XP_016867137.1 5 - 6 812 92
XM_017011649.1 XP_016867138.1 5 - 6 812 92
XM_017011650.1 XP_016867139.1 5 - 6 812 92
XM_017011651.1 XP_016867140.1 5 - 6 812 92
XM_017011652.1 XP_016867141.1 5 - 6 812 92
XM_017011653.1 XP_016867142.1 5 - 6 812 92
XM_017011654.1 XP_016867143.1 5 - 6 812 92
XM_017011655.1 XP_016867144.1 5 - 6 471 92
XM_017011656.1 XP_016867145.1 4 - 5 401 92
GenBank mRNA AB019691.1 - 5 - 6 665 92
AF026245.1 - 6 - 7 799 92
AF247727.4 - 5 - 6 579 92
AJ131693.1 - 5 - 6 801 92
BC015533.2 - 5 - 6 753 92

View Full Product Details