Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
Chromosome Location
Chr.7: 91940867 - 92110673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005751.4 NP_005742.4 6 - 7 961 154
NM_147185.2 NP_671714.1 6 - 7 961 154
XM_017011642.1 XP_016867131.1 6 - 7 969 154
XM_017011643.1 XP_016867132.1 6 - 7 969 154
XM_017011644.1 XP_016867133.1 6 - 7 969 154
XM_017011645.1 XP_016867134.1 5 - 6 915 154
XM_017011646.1 XP_016867135.1 6 - 7 969 154
XM_017011647.1 XP_016867136.1 6 - 7 969 154
XM_017011648.1 XP_016867137.1 6 - 7 969 154
XM_017011649.1 XP_016867138.1 6 - 7 969 154
XM_017011650.1 XP_016867139.1 6 - 7 969 154
XM_017011651.1 XP_016867140.1 6 - 7 969 154
XM_017011652.1 XP_016867141.1 6 - 7 969 154
XM_017011653.1 XP_016867142.1 6 - 7 969 154
XM_017011654.1 XP_016867143.1 6 - 7 969 154
XM_017011655.1 XP_016867144.1 6 - 7 628 154
XM_017011656.1 XP_016867145.1 5 - 6 558 154
GenBank mRNA AB019691.1 - 6 - 7 822 154
AF026245.1 - 7 - 8 956 154
AF247727.4 - 6 - 7 736 154
AJ131693.1 - 6 - 7 958 154
BC015533.2 - 6 - 7 910 154

View Full Product Details