Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGCR2
Entrez Gene ID
9993
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
DGS-C, IDD, LAN, SEZ-12
Chromosome Location
Chr.22: 19036282 - 19122454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173533.1 NP_001167004.1 5 - 6 929 73
NM_001173534.1 NP_001167005.1 5 - 6 920 73
NM_001184781.1 NP_001171710.1 6 - 7 1043 73
NM_005137.2 NP_005128.1 6 - 7 1052 73
NR_033674.1 5 - 6 937 73
GenBank mRNA AK291670.1 - 6 - 7 1010 73
AK295697.1 - 5 - 6 895 73
AK296337.1 - 5 - 6 878 73
AK303130.1 - 6 - 7 962 73
AK304382.1 - 5 - 6 887 73
BC020339.1 - 6 - 7 999 73
BC032430.1 - 6 - 7 988 73
BC035425.1 - 6 - 7 999 73
BC040500.1 - 6 - 7 1047 73
CR456433.1 - 6 - 7 824 73
CR936871.1 - 7 - 8 1387 73
D79985.1 - 6 - 7 989 73
FJ032369.1 - 6 - 7 804 73
L77560.1 - 6 - 7 953 73
X83545.1 - 6 - 7 953 73
X84076.1 - 6 - 7 950 73

View Full Product Details